Homogentisic acid-γ-lactone ≥98.0%

Supplier: TCI America

H0630-1G H0630-5G
TCH0630-001GEA 151.68 CAD
TCH0630-001G TCH0630-005G
Homogentisic acid-γ-lactone ≥98.0%
Homogentisic acid-gamma-lactone

CAS Number: 2688-48-4
MDL Number: MFCD00005857
Molecular Formula: C8H6O3
Molecular Weight: 150.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 193


Other Product Names / Synonyms: 2,3-Dihydro-5-hydroxybenzofuran-2-one, 2,5-Dihydroxyphenylacetic Acid gamma-Lactone, 5-Hydroxy-2-coumaranone

Formula: C₈H₆O₃
Melting Pt: 193 °C
MDL Number: MFCD00005857
CAS Number: 2688-48-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fc2b4afd92dd046f5261a7a4000d93e2ff3749ca&vid=fc2b4afd92dd046f5261a7a4000d93e2ff3749ca&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR