3-Hydroxy-3-methylvaleric acid ≥95.0%

Supplier: TCI America

Synonyms: 3-Hydroxy-3-methylpentanoic Acid

H0626-5G H0626-25G
TCH0626-5GEA 128.93 CAD
TCH0626-5G TCH0626-025G
3-Hydroxy-3-methylvaleric acid ≥95.0%
3-Hydroxy-3-methylvaleric acid

CAS Number: 150-96-9
MDL Number: MFCD00059657
Molecular Formula: C6H12O3
Molecular Weight: 132.16
Purity/Analysis Method: >95.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 144
Specific Gravity (20/20): 1.08


Other Product Names / Synonyms: 3-Hydroxy-3-methylpentanoic Acid

Formula: C₆H₁₂O₃
Boiling Pt: 144 °C
Density: 1.08 (20 °C)
MDL Number: MFCD00059657
CAS Number: 150-96-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13a0ea2319689892800b36c57c32f4405e34ba45&vid=13a0ea2319689892800b36c57c32f4405e34ba45&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR