2-Quinoxalinol ≥98.0%
Supplier: TCI America
Synonyms:
2-Quinoxalinone, 2(1H)-Quinoxalinone, 2-Hydroxyquinoxaline, quinoxalin-2-ol
H0591-25G
H0591-500G
H0591-100G
TCH0591-025GEA
46.29
CAD
TCH0591-025G
TCH0591-500G
TCH0591-100G
2-Quinoxalinol ≥98.0%
2-Quinoxalinol
CAS Number: 1196-57-2
MDL Number: MFCD00006722
Molecular Formula: C8H6N2O
Molecular Weight: 146.15
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 275
Other Product Names / Synonyms: 2-Quinoxalinol
Formula:
C₈H₆N₂O MW: 146.15 g/mol Melting Pt: 275 °C |
MDL Number:
MFCD00006722 CAS Number: 1196-57-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...