N-Allylrhodanine ≥98.0%

Supplier: TCI America

A1412-5G A1412-25G
TCA1412-005GEA 131.14 CAD
TCA1412-005G TCA1412-025G
N-Allylrhodanine ≥98.0%
N-Allylrhodanine

CAS Number: 1457-47-2
MDL Number: MFCD00044700
Molecular Formula: C6H7NOS2
Molecular Weight: 173.25
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 139
Melting point (°C): 47


Other Product Names / Synonyms: 3-Allyl-2-thioxo-4-thiazolidinone

Formula: C₆H₇NOS₂
Boiling Pt: 139 °C
Melting Pt: 47 °C
MDL Number: MFCD00044700
CAS Number: 1457-47-2
EINECS: 215-941-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=828700c7b90fb113474a5f480afff680bba47c8c&vid=828700c7b90fb113474a5f480afff680bba47c8c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR