2-(2-Hydroxy-5-methylphenyl)benzotriazole ≥99.0%

Supplier: TCI America

Synonyms: 2-(2-Benzotriazolyl)-p-cresol, Drometrizole

H0560-25G H0560-500G
TCH0560-025GEA 119.42 CAD
TCH0560-025G TCH0560-500G
2-(2-Hydroxy-5-methylphenyl)benzotriazole ≥99.0%
2-(2-Hydroxy-5-methylphenyl)benzotriazole

CAS Number: 2440-22-4
MDL Number: MFCD00022903
Molecular Formula: C13H11N3O
Molecular Weight: 225.25
Purity/Analysis Method: >99.0% (HPLC,T)
Form: Crystal
Melting point (°C): 130
Flash Point (°C): 210


Product Categories: Materials Chemistry, Resin Additives
Other Product Names / Synonyms: 2-(2-Benzotriazolyl)-p-cresol

Formula: C₁₃H₁₁N₃O
Melting Pt: 130 °C
Flash Pt: 210 °C
MDL Number: MFCD00022903
CAS Number: 2440-22-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd426bebe5ffc6543621643d7560b3fef622fc16&vid=fd426bebe5ffc6543621643d7560b3fef622fc16&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR