2-(5-Chloro-2-benzotriazolyl)-6-tert-butyl-p-cresol ≥98.0%
Supplier: TCI America
H0559-25G
H0559-500G
TCH0559-025GEA
109.32
CAD
TCH0559-025G
TCH0559-500G
2-(5-Chloro-2-benzotriazolyl)-6-tert-butyl-p-cresol ≥98.0%
2-(5-Chloro-2-benzotriazolyl)-6-tert-butyl-p-cresol
CAS Number: 3896-11-5
MDL Number: MFCD00059707
Molecular Formula: C17H18ClN3O
Molecular Weight: 315.80
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 140
Product Categories: Materials Chemistry, Resin Additives
Other Product Names / Synonyms: 2-(2-Hydroxy-3-tert-butyl-5-methylphenyl)-5-chlorobenzotriazole
Formula:
C₁₇H₁₈ClN₃O Melting Pt: 140 °C |
MDL Number:
MFCD00059707 CAS Number: 3896-11-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...