Nickel(II) hexafluoroacetylacetonate hydrate

Supplier: TCI America

Synonyms: Hexafluoroacetylacetono Nickel(II) Salt, Bis(hexafluoroacetylacetonato)nickel(II) hydrate

H0558-1G H0558-5G
TCH0558-001GEA 134.3 CAD
TCH0558-001G TCH0558-5G
Nickel(II) hexafluoroacetylacetonate hydrate
Nickel(II) hexafluoroacetylacetonate hydrate

Color: Pale Yellow


Other Product Names / Synonyms: Hexafluoroacetylacetono Nickel(II) Salt, Nickel(II) Hexafluoroacetylacetonate

Formula: C₁₀H₂F₁₂NiO₄·xH₂O
MDL Number: MFCD00067506
CAS Number: 14949-69-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=10f021262be65f0838c79cc5bafd2b5f7d659e90&vid=10f021262be65f0838c79cc5bafd2b5f7d659e90&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR