Bis(hexafluoroacetylacetonato)manganese(II) hydrate ≥95.0% (by titrimetric analysis)

Supplier: TCI America

H0557-5G H0557-1G
TCH0557-5GEA 298.62 CAD
TCH0557-5G TCH0557-001G
Bis(hexafluoroacetylacetonato)manganese(II) hydrate ≥95.0% (by titrimetric analysis)
Bis(hexafluoroacetylacetonato)manganese(II) hydrate

CAS Number: 19648-86-3
MDL Number: MFCD00042513
Molecular Formula: C10H2F12MnO4
Molecular Weight: 469.04
Purity/Analysis Method: >95.0% (T)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Materials Chemistry, Magnetic Metal Complexes (Magnetic Materials)
Other Product Names / Synonyms: Hexafluoroacetylacetono Manganese(II) Salt, Manganese(II) Hexafluoroacetylacetonate

Formula: C₁₀H₂F₁₂MnO₄
MDL Number: MFCD00042513
CAS Number: 19648-86-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2197bb3c9048d49c02937818a4239442f94fa1f5&vid=c6d631ef51a89863703fe07b2ad40958588d2df4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR