Copper(II) hexafluoroacetylacetonate hydrate ≥95.0%

Supplier: TCI America

Synonyms: Hexafluoroacetylacetono Copper(II) Salt, Copper(II) Hexafluoroacetylacetonate, Copper(II) hexafluoro-2,4-pentanedionate hydrate, Bis(hexafluoroacetylacetonato)copper(II) hydrate, Copper(II) hexafluoro-2,4-pentanedionate xhydrate

H0554-1G H0554-5G
TCH0554-001GEA 93.22 CAD
TCH0554-001G TCH0554-5G
Copper(II) hexafluoroacetylacetonate hydrate ≥95.0%
Copper (II) hexafluoroacetylacetonate hydrate

CAS Number: 14781-45-4
MDL Number: MFCD00064754
Molecular Formula: C10H2CuF12O4
Molecular Weight: 477.65
Purity/Analysis Method: >95.0% (T)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Materials Chemistry, Magnetic Metal Complexes (Magnetic Materials)
Other Product Names / Synonyms: Copper(II) Hexafluoroacetylacetonate, Hexafluoroacetylacetono Copper(II) Salt

Formula: C₁₀H₂CuF₁₂O₄
MW: 477.64 g/mol (anhy)
MDL Number: MFCD00064754
CAS Number: 14781-45-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6300729a5401cc64b6484c4d937aded5960636d8&vid=6300729a5401cc64b6484c4d937aded5960636d8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR