Methyl-5-aminolevulinate hydrochloride ≥98.0%
Supplier: TCI America
Synonyms:
Methyl aminolevulinate hydrochloride, δ-Aminolevulinic Acid Methyl Ester Hydrochloride, 3-Aminoacetylpropionic Acid Methyl Ester Hydrochloride, 5-Amino-4-oxovaleric Acid Methyl Ester Hydrochloride, Methyl 5-amino-4-oxopentanoate hydrochloride, 5-Aminolevulinic Acid Methyl Ester Hydrochloride
CAS Number: 79416-27-6
MDL Number: MFCD00012868
Molecular Formula: C6H11NO3
Molecular Weight: 181.62
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal
Melting point (°C): 122
Storage Temperature: 0-10°C
Other Product Names / Synonyms: 3-Aminoacetylpropionic Acid Methyl Ester Hydrochloride, 5-Aminolevulinic Acid Methyl Ester Hydrochloride, 5-Amino-4-oxovaleric Acid Methyl Ester Hydrochloride
Formula:
C₆H₁₁NO₃ MW: 181.62 g/mol Melting Pt: 122 °C |
MDL Number:
MFCD00012868 CAS Number: 79416-27-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...