1-(3-Hydroxypropyl)-2-pyrrolidone ≥96.0%
Supplier: TCI America
H0541-10ML
H0541-25ML
TCH0541-010MLEA
91.64
CAD
TCH0541-010ML
TCH0541-25ML
1-(3-Hydroxypropyl)-2-pyrrolidone ≥96.0%
1-(3-Hydroxypropyl)-2-pyrrolidone
CAS Number: 62012-15-1
MDL Number: MFCD00059014
Molecular Formula: C7H13NO2
Molecular Weight: 143.19
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 140
Flash Point (°C): 110
Specific Gravity (20/20): 1.11
Formula:
C₇H₁₃NO₂ Boiling Pt: 140 °C Density: 1.11 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00059014 CAS Number: 62012-15-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...