Ethyl heptadecanoate ≥97.0%

Supplier: TCI America

H0526-5G H0526-25G
TCH0526-5GEA 161.16 CAD
TCH0526-5G TCH0526-025G
Ethyl heptadecanoate ≥97.0%
Ethyl heptadecanoate

CAS Number: 14010-23-2
MDL Number: MFCD00026658
Molecular Formula: C19H38O2
Molecular Weight: 298.51
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 200
Freezing point (°C): 28


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Ethyl Margarate, Heptadecanoic Acid Ethyl Ester, Margaric Acid Ethyl Ester

Formula: C₁₉H₃₈O₂
Boiling Pt: 200 °C
MDL Number: MFCD00026658
CAS Number: 14010-23-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e5bcf27a5b0ee2d7e37c1486aef31bc8c5963c9e&vid=e5bcf27a5b0ee2d7e37c1486aef31bc8c5963c9e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR