(±)-1-Indanol ≥99.0%

Supplier: TCI America

H0516-5G H0516-25G
TCH0516-005GEA 56.88 CAD
TCH0516-005G TCH0516-025G
(±)-1-Indanol ≥99.0%
(±)-1-Indanol

CAS Number: 6351-10-6
MDL Number: MFCD00003797
Molecular Formula: C9H10O
Molecular Weight: 134.18
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 220
Melting point (°C): 52
Flash Point (°C): 145


Other Product Names / Synonyms: 1-Hydroxyhydrindene, 1-Indanol

Formula: C₉H₁₀O
Boiling Pt: 220 °C
Melting Pt: 52 °C
Flash Pt: 145 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003797
CAS Number: 6351-10-6
EINECS: 228-755-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b838007a37cbc281dcbb1a0a2fc24b888351b74d&vid=20f58553421f33b21aa5b1c38f5a313cfe87298f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR