4-(Hydroxyamino)quinoline-N-oxide ≥97.0%

Supplier: TCI America

Synonyms: N-(4-Quinolyl)hydroxylamine 1'-Oxide

H0515-1G H0515-5G
TCH0515-001GEA 221.2 CAD
TCH0515-001G TCH0515-5G
4-(Hydroxyamino)quinoline-N-oxide ≥97.0%
4-(Hydroxyamino)quinoline-N-oxide

CAS Number: 4637-56-3
MDL Number: MFCD00059752
Molecular Formula: C9H8N2O2
Molecular Weight: 176.18
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 214


Other Product Names / Synonyms: N-(4-Quinolyl)hydroxylamine 1'-Oxide

Formula: C₉H₈N₂O₂
MW: 176.18 g/mol
Melting Pt: 214 °C
MDL Number: MFCD00059752
CAS Number: 4637-56-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=04161cc8778c47e2cc95e94a2f7323ac945b0086&vid=04161cc8778c47e2cc95e94a2f7323ac945b0086&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR