5-Hydroxy-2-pentanone ≥96.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 5-Hydroxypentan-2-one, 3-Acetyl-1-propanol

H0511-250ML H0511-25ML
TCH0511-250MLEA 492.96 CAD
TCH0511-250ML TCH0511-025ML
5-Hydroxy-2-pentanone ≥96.0% (by titrimetric analysis)
5-Hydroxy-2-pentanone

CAS Number: 1071-73-4
MDL Number: MFCD00002961
Molecular Formula: C5H10O2
Molecular Weight: 102.13
Purity/Analysis Method: >96.0% (T)
Form: Clear Liquid
Boiling point (°C): 118
Flash Point (°C): 93
Specific Gravity (20/20): 1.00


Other Product Names / Synonyms: 3-Acetyl-1-propanol

Formula: C₅H₁₀O₂
MW: 102.13 g/mol
Boiling Pt: 118 °C
Density: 1.00 (20 °C)
Flash Pt: 93 °C
MDL Number: MFCD00002961
CAS Number: 1071-73-4
EINECS: 213-994-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9029cb1117336296576925e279671c85c30d6283&vid=9029cb1117336296576925e279671c85c30d6283&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR