2-Ethylhexyl-4-hydroxybenzoate ≥98.0%

Supplier: TCI America

Synonyms: 4-Hydroxybenzoic Acid Octyl Ester, 4-Hydroxybenzoic Acid 2-Ethylhexyl Ester, Octyl 4-Hydroxybenzoate, 2-Ethylhexylparaben, Octylparaben, 2-Ethylhexyl 4-hydroxybenzoate

H0506-500G H0506-25G
TCH0506-500GEA 195.92 CAD
TCH0506-500G TCH0506-025G
2-Ethylhexyl-4-hydroxybenzoate ≥98.0%
2-Ethylhexyl-4-hydroxybenzoate

CAS Number: 5153-25-3
MDL Number: MFCD00016472
Molecular Formula: C15H22O3
Molecular Weight: 250.34
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Clear Liquid
Boiling point (°C): 270
Specific Gravity (20/20): 1.04


Other Product Names / Synonyms: 2-Ethylhexylparaben, 4-Hydroxybenzoic Acid 2-Ethylhexyl Ester, 4-Hydroxybenzoic Acid Octyl Ester, Octyl 4-Hydroxybenzoate, Octylparaben

Formula: C₁₅H₂₂O₃
Boiling Pt: 270 °C
Density: 1.04 (20 °C)
MDL Number: MFCD00016472
CAS Number: 5153-25-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2fc6aaca10b8647460efb2911f46d0f20402fe9&vid=e2fc6aaca10b8647460efb2911f46d0f20402fe9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR