sec-Butyl-4-hydroxybenzoate ≥98.0%

Supplier: TCI America

Synonyms: 4-Hydroxybenzoic acid sec-butyl ester

H0496-500G H0496-25G H0496-100G
TCH0496-500GEA 1422 CAD
TCH0496-500G TCH0496-025G TCH0496-100G
sec-Butyl-4-hydroxybenzoate ≥98.0%
sec-Butyl 4-hydroxybenzoate

CAS Number: 17696-61-6
MDL Number: MFCD00059626
Molecular Formula: C11H14O3
Molecular Weight: 194.23
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Boiling point (°C): 210
Melting point (°C): 60


Other Product Names / Synonyms: 4-Hydroxybenzoic Acid sec-Butyl Ester, sec-Butylparaben

Formula: C₁₁H₁₄O₃
Boiling Pt: 210 °C
Melting Pt: 60 °C
MDL Number: MFCD00059626
CAS Number: 17696-61-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea33d502a51364e9dc566bc26f3a25ece89d2c8f&vid=ea33d502a51364e9dc566bc26f3a25ece89d2c8f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR