Isobutyl-4-hydroxybenzoate ≥99.0% (by HPLC, titration analysis)

Supplier: TCI America

H0493-500G H0493-25G H0493-100G
TCH0493-500GEA 401.32 CAD
TCH0493-500G TCH0493-025G TCH0493-100G
Isobutyl-4-hydroxybenzoate ≥99.0% (by HPLC, titration analysis)
Isobutyl 4-hydroxybenzoate

CAS Number: 4247-02-3
MDL Number: MFCD00020167
Molecular Formula: C11H14O3
Molecular Weight: 194.23
Purity/Analysis Method: >99.0% (HPLC,T)
Form: Crystal
Melting point (°C): 76


Other Product Names / Synonyms: 4-Hydroxybenzoic Acid Isobutyl Ester, Isobutylparaben

Formula: C₁₁H₁₄O₃
Melting Pt: 76 °C
MDL Number: MFCD00020167
CAS Number: 4247-02-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1cd9f47d0da18d8ca43c2b02bbb51d7a36530c28&vid=1cd9f47d0da18d8ca43c2b02bbb51d7a36530c28&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR