3-Hydroxypyridine-N-oxide ≥96.0%
Supplier: TCI America
Synonyms:
3-Pyridinol N-Oxide, 3-Hydroxypyridine 1-oxide
H0492-5G
H0492-25G
TCH0492-5GEA
64.78
CAD
TCH0492-5G
TCH0492-025G
3-Hydroxypyridine-N-oxide ≥96.0%
3-Hydroxypyridine N-oxide
CAS Number: 6602-28-4
MDL Number: MFCD00006200
Molecular Formula: C5H5NO2
Molecular Weight: 111.10
Purity/Analysis Method: >96.0% (GC,T)
Form: Crystal
Melting point (°C): 190
Other Product Names / Synonyms: 3-Pyridinol N-Oxide
Formula:
C₅H₅NO₂ MW: 111.0987 g/mol Melting Pt: 190 °C |
MDL Number:
MFCD00006200 CAS Number: 6602-28-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...