N-(2-Hydroxyethyl)iminodiacetic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
H0489-25G
H0489-5G
TCH0489-025GEA
238.58
CAD
TCH0489-025G
TCH0489-005G
N-(2-Hydroxyethyl)iminodiacetic acid ≥98.0% (by titrimetric analysis)
N-(2-Hydroxyethyl)iminodiacetic acid
CAS Number: 93-62-9
MDL Number: MFCD00004293
Molecular Formula: C6H11NO5
Molecular Weight: 177.16
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 177
Other Product Names / Synonyms: N,N-Bis(carboxymethyl)ethanolamine
Formula:
C₆H₁₁NO₅ Melting Pt: 177 °C |
MDL Number:
MFCD00004293 CAS Number: 93-62-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...