trans-3-Hexenedioic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: trans-beta-Hydromuconic Acid, trans-2-Butene-1,4-dicarboxylic Acid, Dihydromuconic Acid, Hex-3-enedioic acid

H0477-5G H0477-25G
TCH0477-005GEA 159.17 CAD
TCH0477-005G TCH0477-025G
trans-3-Hexenedioic acid ≥98.0% (by titrimetric analysis)
trans-3-Hexenedioic acid

CAS Number: 4436-74-2
MDL Number: MFCD00002785
Molecular Formula: C6H8O4
Molecular Weight: 144.13
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 198


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: trans-2-Butene-1,4-dicarboxylic Acid, Dihydromuconic Acid, trans-beta-Hydromuconic Acid

Formula: C₆H₈O₄
MW: 144.13 g/mol
Melting Pt: 198 °C
MDL Number: MFCD00002785
CAS Number: 4436-74-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f947321a79f4832582afc70d6b2f7d038b06b5c4&vid=f947321a79f4832582afc70d6b2f7d038b06b5c4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR