2-Hexadecenoic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

H0428-1G H0428-5G
TCH0428-001GEA 303.36 CAD
TCH0428-001G TCH0428-005G
2-Hexadecenoic acid ≥98.0% (by titrimetric analysis)
2-Hexadecenoic acid

CAS Number: 629-56-1
MDL Number: MFCD00045907
Molecular Formula: C16H30O2
Molecular Weight: 254.41
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 49


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Gaidic Acid

Formula: C₁₆H₃₀O₂
Melting Pt: 49 °C
MDL Number: MFCD00045907
CAS Number: 629-56-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b8e1a2184e75d8a49c48ce7de16c3a3d62410250&vid=b8e1a2184e75d8a49c48ce7de16c3a3d62410250&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR