Hydroxyethyl cellulose

Supplier: TCI America

Synonyms: Hydroxyethyl Cellulose

H0418-25G H0418-500G
TCH0418-25GEA 47.4 CAD
TCH0418-25G TCH0418-500G
Hydroxyethyl cellulose
Hydroxyethyl cellulose

Form: Crystal
Color: Very Pale Yellow


Product Categories: Materials Chemistry, Polymers, Bioscience, Polysaccharides, Cellulose, Natural Polymers (for DDS)

MDL Number: MFCD00072770
CAS Number: 9004-62-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR