1-(2-Hydroxyethyl)-2-pyrrolidone ≥98.0%

Supplier: TCI America

Synonyms: N-(2-Hydroxyethyl)-2-pyrrolidone

H0408-25G H0408-500G
TCH0408-025GEA 44.24 CAD
TCH0408-025G TCH0408-500G
1-(2-Hydroxyethyl)-2-pyrrolidone ≥98.0%
1-(2-Hydroxyethyl)-2-pyrrolidone

CAS Number: 3445-11-2
MDL Number: MFCD00014103
Molecular Formula: C6H11NO2
Molecular Weight: 129.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 175
Flash Point (°C): 174
Freezing point (°C): 25

Formula: C₆H₁₁NO₂
MW: 129.16 g/mol
Boiling Pt: 175 °C
Flash Pt: 174 °C
MDL Number: MFCD00014103
CAS Number: 3445-11-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=79fed451aa3a007441ac5de0f2e1a2ed793bf4a1&vid=79fed451aa3a007441ac5de0f2e1a2ed793bf4a1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR