Hydroxyethyl cellulose
Supplier: TCI America
Synonyms:
Hydroxyethyl Cellulose
H0392-25G
H0392-500G
TCH0392-25GEA
47.4
CAD
TCH0392-25G
TCH0392-500G
Hydroxyethyl cellulose
Hydroxyethyl cellulose
Form: Crystal
Color: White
Product Categories: Materials Chemistry, Polymers, Bioscience, Polysaccharides, Cellulose, Natural Polymers (for DDS)
MDL Number:
MFCD00072770 CAS Number: 9004-62-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...