Hydroxydicyclopentadiene ≥90.0% (so called)
Supplier: TCI America
Synonyms:
8(9)-Hydroxytricyclo[5.2.1.0(2,6)]dec-3-ene
H0384-25ML
H0384-500ML
TCH0384-025MLEA
50.56
CAD
TCH0384-025ML
TCH0384-500ML
Hydroxydicyclopentadiene ≥90.0% (so called)
Hydroxydicyclopentadiene
CAS Number: 37275-49-3
MDL Number: MFCD00151152
Molecular Formula: C10H14O
Molecular Weight: 150.22
Purity/Analysis Method: >90.0% (GC)
Form: Viscous Clear Liquid
Flash Point (°C): 106
Specific Gravity (20/20): 1.08
Other Product Names / Synonyms: 8(9)-Hydroxytricyclo[5.2.1.0(2,6)]dec-3-ene
Formula:
C₁₀H₁₄O MW: 150.22 g/mol Density: 1.08 (20 °C) Flash Pt: 106 °C |
MDL Number:
MFCD00151152 CAS Number: 37275-49-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...