Heptyl sulfide ≥97.0%

Supplier: TCI America

Synonyms: Diheptyl Sulfide, Diheptyl Sulphide

H0376-25G H0376-5G
TCH0376-25GEA 687.3 CAD
TCH0376-25G TCH0376-5G
Heptyl sulfide ≥97.0%
Heptyl sulfide

CAS Number: 629-65-2
MDL Number: MFCD00027315
Molecular Formula: C14H30S
Molecular Weight: 230.45
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 298
Melting point (°C): -11
Specific Gravity (20/20): 0.84


Other Product Names / Synonyms: Diheptyl Sulfide

Formula: C₁₄H₃₀S
MW: 230.45 g/mol
Boiling Pt: 298 °C
Melting Pt: –11 °C
Density: 0.84 (20 °C)
MDL Number: MFCD00027315
CAS Number: 629-65-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d7a7ab0462d0625d95154b0ed22a547951ae975&vid=7d7a7ab0462d0625d95154b0ed22a547951ae975&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR