5-Hydroxyindol-3-ylacetic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

H0355-1G H0355-100MG
TCH0355-001GEA 592.5 CAD
TCH0355-001G TCH0355-100MG
5-Hydroxyindol-3-ylacetic acid ≥98.0% (by HPLC, titration analysis)
5-Hydroxyindol-3-ylacetic acid

CAS Number: 54-16-0
MDL Number: MFCD00005639
Molecular Formula: C10H9NO3
Molecular Weight: 191.19
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 162

Formula: C₁₀H₉NO₃
Melting Pt: 162 °C
Storage Temperature: Freezer
MDL Number: MFCD00005639
CAS Number: 54-16-0
EINECS: 200-195-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e6f7fa34bcde10f06db67b0b198e6ce4d7193efe&vid=e6f7fa34bcde10f06db67b0b198e6ce4d7193efe&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR