1-Hydroxy-9,10-anthraquinone ≥98.0%

Supplier: TCI America

Synonyms: 1-Hydroxyanthraquinone

H0354-25G H0354-5G
TCH0354-025GEA 271.76 CAD
TCH0354-025G 76220-106
1-Hydroxy-9,10-anthraquinone ≥98.0%
1-Hydroxy-9,10-anthraquinone

CAS Number: 129-43-1 MDL Number: MFCD00058946 Molecular Formula: C14H8O3 Molecular Weight: 224.22 Purity/Analysis Method: 98.0% (HPLC) Form: Crystal Melting point (°C): 198

Formula: C₁₄H₈O₃
MW: 224.2115 g/mol
Melting Pt: 198 °C
MDL Number: MFCD00058946
CAS Number: 129-43-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7efd9bde1b8ac776f6153c8186962eb6710cb4d&vid=a7efd9bde1b8ac776f6153c8186962eb6710cb4d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR