Sodium-4-hydroxyazobenzene-4'-sulfonate hydrate ≥98.0%

Supplier: TCI America

Synonyms: 4-Hydroxyazobenzene-4'-sulfonic acid Sodium Salt, 4-Hydroxyazobenzene-4'-sulphonic acid Sodium Salt

H0327-25G H0327-5G
TCH0327-025GEA 350.44 CAD
TCH0327-025G TCH0327-5G
Sodium-4-hydroxyazobenzene-4'-sulfonate hydrate ≥98.0%
Sodium-4-hydroxyazobenzene-4'-sulfonate hydrate

CAS Number: 2623-36-1
MDL Number: MFCD00068861
Molecular Formula: C12H10N2O4S
Molecular Weight: 300.26
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Other Product Names / Synonyms: 4-Hydroxyazobenzene-4'-sulfonic Acid Sodium Salt

Formula: C₁₂H₁₀N₂O₄S
MDL Number: MFCD00068861
CAS Number: 2623-36-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=965274dd4577348c77d6af59079269f96cafa4e5&vid=965274dd4577348c77d6af59079269f96cafa4e5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR