8-Hydroxyquinoline-5-sulfonic acid monohydrate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

H0306-500G H0306-25G
TCH0306-500GEA 1104.1 CAD
TCH0306-500G TCH0306-025G
8-Hydroxyquinoline-5-sulfonic acid monohydrate ≥98.0% (by titrimetric analysis)
8-Hydroxyquinoline-5-sulphonic acid monohydrate

CAS Number: 283158-18-9
MDL Number: MFCD00006795
Molecular Formula: C9H7NO4S
Molecular Weight: 225.22
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 314


Other Product Names / Synonyms: Oxine-5-sulfonic Acid, 8-Quinolinol-5-sulfonic Acid, 8-Quinolinone-5-sulfonic Acid

Formula: C₉H₇NO₄S·1H₂O
Melting Pt: 314 °C
MDL Number: MFCD00149482
CAS Number: 283158-18-9
EINECS: 201-570-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a28ca40ee2c46ec1c6ec556d42f63f9817b8feab&vid=a28ca40ee2c46ec1c6ec556d42f63f9817b8feab&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR