4-(1-Adamantyl)phenol ≥99.0%

Supplier: TCI America

A1392-5G A1392-1G
TCA1392-005GEA 205.4 CAD
TCA1392-005G TCA1392-001G
4-(1-Adamantyl)phenol ≥99.0%
4-(1-Adamantyl)phenol

CAS Number: 29799-07-3
MDL Number: MFCD00168143
Molecular Formula: C16H20O
Molecular Weight: 228.34
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 179


Other Product Names / Synonyms: 1-(4-Hydroxyphenyl)adamantane

Formula: C₁₆H₂₀O
Melting Pt: 179 °C
MDL Number: MFCD00168143
CAS Number: 29799-07-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5a0fd28e3dc2d4661d3ec99090336ffa9b75cfda&vid=5a0fd28e3dc2d4661d3ec99090336ffa9b75cfda&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR