4-Hydroxyphenylpyruvic acid ≥95.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
3-(4-Hydroxyphenyl)-2-oxopropanoic acid
H0294-5G
H0294-1G
TCH0294-005GEA
537.2
CAD
TCH0294-005G
TCH0294-001G
4-Hydroxyphenylpyruvic acid ≥95.0% (by HPLC, titration analysis)
4-Hydroxyphenylpyruvic acid
CAS Number: 156-39-8
MDL Number: MFCD00002591
Molecular Formula: C9H8O4
Molecular Weight: 180.16
Purity/Analysis Method: >95.0% (HPLC,T)
Form: Crystal
Storage Temperature: 0-10°C
Formula:
C₉H₈O₄ MW: 180.16 g/mol |
MDL Number:
MFCD00002591 CAS Number: 156-39-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...