2-(2-Hydroxyphenyl)benzoxazole ≥98.0% (by GC, titration analysis)
Supplier: TCI America
H0291-1G
H0291-25G
TCH0291-001GEA
47.4
CAD
TCH0291-001G
TCH0291-025G
2-(2-Hydroxyphenyl)benzoxazole ≥98.0% (by GC, titration analysis)
2-(2-Hydroxyphenyl)benzoxazole
CAS Number: 835-64-3
MDL Number: MFCD00005767
Molecular Formula: C13H9NO2
Molecular Weight: 211.22
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 338
Melting point (°C): 124
Formula:
C₁₃H₉NO₂ Boiling Pt: 338 °C Melting Pt: 124 °C |
MDL Number:
MFCD00005767 CAS Number: 835-64-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...