β-Alaninamide hydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 3-Aminopropionamide Hydrochloride, H-β-Ala-NH₂·HCl, H-β-Ala-NH2.HCl, H-β-Ala-NH2·HCl

A1391-1G A1391-5G
TCA1391-001GEA 282.82 CAD
TCA1391-001G TCA1391-5G
β-Alaninamide hydrochloride ≥98.0% (by titrimetric analysis)
beta-Alaninamide hydrochloride

CAS Number: 64017-81-8
MDL Number: MFCD00060747
Molecular Formula: C3H8N2O
Molecular Weight: 124.57
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 153


Product Categories: Bioscience, Amino Acid Amides
Other Product Names / Synonyms: 3-Aminopropionamide Hydrochloride

Formula: C₃H₈N₂O·HCl
MW: 124.57 g/mol
Melting Pt: 153 °C
MDL Number: MFCD00060747
CAS Number: 64017-81-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0783ced91991baa1982653027a3d54fc0df83b84&vid=0783ced91991baa1982653027a3d54fc0df83b84&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR