3-Hydroxymyristic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

H0274-1G H0274-5G
TCH0274-1GEA 124.82 CAD
TCH0274-1G TCH0274-005G
3-Hydroxymyristic acid ≥98.0% (by GC, titration analysis)
3-Hydroxymyristic acid

CAS Number: 1961-72-4
MDL Number: MFCD00059633
Molecular Formula: C14H28O3
Molecular Weight: 244.38
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 80


Other Product Names / Synonyms: 3-Hydroxytetradecanoic Acid

Formula: C₁₄H₂₈O₃
Melting Pt: 80 °C
MDL Number: MFCD00059633
CAS Number: 1961-72-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=59b1619b1b837dd2947740f23644bda755e71eb6&vid=59b1619b1b837dd2947740f23644bda755e71eb6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR