N-(2-Hydroxyethyl)ethylenediamine-N,N',N'-triacetic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

H0243-500G H0243-25G H0243-100G
TCH0243-500GEA 371.3 CAD
TCH0243-500G TCH0243-025G TCH0243-100G
N-(2-Hydroxyethyl)ethylenediamine-N,N',N'-triacetic acid ≥98.0% (by titrimetric analysis)
N-(2-Hydroxyethyl)ethylenediamine-N,N',N'-triacetic acid

CAS Number: 150-39-0
MDL Number: MFCD00004294
Molecular Formula: C10H18N2O7
Molecular Weight: 278.26
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Product Categories: Analytical Science, Complexones (Chelating Agents)
Other Product Names / Synonyms: N-Carboxymethyl-N'-(2-hydroxyethyl)-N,N'-ethylenediglycine, Versenol, HEDTA

Formula: C₁₀H₁₈N₂O₇
MDL Number: MFCD00004294
CAS Number: 150-39-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=309f5cce548032235b555e92744ddc16c0ed871e&vid=309f5cce548032235b555e92744ddc16c0ed871e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR