4-Hydroxy-4'-dimethylaminoazobenzene ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
H0237-25G
H0237-1G
TCH0237-025GEA
511.92
CAD
TCH0237-025G
TCH0237-001G
4-Hydroxy-4'-dimethylaminoazobenzene ≥98.0% (by HPLC, titration analysis)
4-Hydroxy-4'-dimethylaminoazobenzene
CAS Number: 2496-15-3
MDL Number: MFCD00059624
Molecular Formula: C14H15N3O
Molecular Weight: 241.29
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 200
Other Product Names / Synonyms: 4-(4-Dimethylaminophenylazo)phenol
Formula:
C₁₄H₁₅N₃O Melting Pt: 200 °C |
MDL Number:
MFCD00059624 CAS Number: 2496-15-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...