4-Hydroxybutyl acrylate ≥97.0% stabilised

Supplier: TCI America

Synonyms: Acrylic Acid 4-Hydroxybutyl Ester

A1390-25G A1390-500G A1390-100G
TCA1390-025GEA 81.05 CAD
TCA1390-025G TCA1390-500G TCA1390-100G
4-Hydroxybutyl acrylate ≥97.0% stabilised
4-Hydroxybutyl acrylate

(stabilized with MEHQ)
CAS Number: 2478-10-6
MDL Number: MFCD00010261
Molecular Formula: C7H12O3
Molecular Weight: 144.17
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 95
Flash Point (°C): 113
Specific Gravity (20/20): 1.04


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Acrylic Acid 4-Hydroxybutyl Ester

Formula: C₇H₁₂O₃
Boiling Pt: 95 °C
Density: 1.04 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00010261
CAS Number: 2478-10-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=61cf78363a39ac50322eb09ad6354d86864bd6c1&vid=61cf78363a39ac50322eb09ad6354d86864bd6c1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR