Ethyl-3-hydroxybutyrate ≥98.0%
Supplier: TCI America
H0230-500G
H0230-25G
TCH0230-500GEA
312.84
CAD
TCH0230-500G
TCH0230-025G
Ethyl-3-hydroxybutyrate ≥98.0%
Ethyl 3-hydroxybutyrate
CAS Number: 5405-41-4
MDL Number: MFCD00004545
Molecular Formula: C6H12O3
Molecular Weight: 132.16
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 180
Flash Point (°C): 64
Specific Gravity (20/20): 1.02
Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: DL-3-Hydroxybutyric Acid Ethyl Ester
Formula:
C₆H₁₂O₃ Boiling Pt: 180 °C Density: 1.02 (20 °C) Flash Pt: 64 °C |
MDL Number:
MFCD00004545 CAS Number: 5405-41-4 EINECS: 226-456-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...