Ethyl-2-hydroxybutyrate ≥95.0%

Supplier: TCI America

H0229-5G H0229-25G
TCH0229-005GEA 494.54 CAD
TCH0229-005G TCH0229-25G
Ethyl-2-hydroxybutyrate ≥95.0%
Ethyl-2-hydroxybutyrate

CAS Number: 52089-54-0
MDL Number: MFCD00014407
Molecular Formula: C6H12O3
Molecular Weight: 132.16
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 167
Specific Gravity (20/20): 1.01


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: 2-Hydroxybutyric Acid Ethyl Ester

Formula: C₆H₁₂O₃
Boiling Pt: 167 °C
Density: 1.01 (20 °C)
MDL Number: MFCD00014407
CAS Number: 52089-54-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=021d072aeb070075d03a1f305602486ce9d2dd5e&vid=021d072aeb070075d03a1f305602486ce9d2dd5e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR