(±)-3-Hydroxybutyric acid ≥75.0% (by titrimetric analysis) (contains Polymolecular esterification product)

Supplier: TCI America
Danger

Synonyms: 3-Hydroxybutyric acid, DL-3-Hydroxybutyric Acid, (±)-3-Hydroxybutanoic acid

H0228-25G H0228-5G
TCH0228-025GEA 409.22 CAD
TCH0228-025G TCH0228-005G
(±)-3-Hydroxybutyric acid ≥75.0% (by titrimetric analysis) (contains Polymolecular esterification product)
(±)-3-Hydroxybutyric acid

CAS Number: 300-85-6
MDL Number: MFCD00004546
Molecular Formula: C4H8O3
Molecular Weight: 104.11
Purity/Analysis Method: >75.0% (T)
Form: Clear Liquid
Flash Point (°C): 110
Storage Temperature: 0-10°C

Formula: C₄H₈O₃
MW: 104.11 g/mol
Boiling Pt: 269 °C (1013 hPa)
Melting Pt: 33…34 °C
Density: 1.26 g/cm³ (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00004546
CAS Number: 300-85-6
EINECS: 206-099-9
Merck Index: 12,04861
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d7a0a8f9d154510de060f84ad32bde5cf9f9558&vid=6d7a0a8f9d154510de060f84ad32bde5cf9f9558&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 75.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR