Salicyl alcohol ≥98.0%
Supplier: TCI America
H0223-250G
H0223-25G
TCH0223-250GEA
1093.36
CAD
TCH0223-250G
TCH0223-025G
Salicyl alcohol ≥98.0%
Salicyl alcohol
CAS Number: 90-01-7
MDL Number: MFCD00004617
Molecular Formula: C7H8O2
Molecular Weight: 124.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 86
Other Product Names / Synonyms: 2-Methylolphenol, Salicyl Alcohol, Saligenin
Formula:
C₇H₈O₂ Boiling Pt: 268 °C (1013 hPa) Melting Pt: 86 °C Density: 1.1613 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004617 CAS Number: 90-01-7 EINECS: 201-960-5 Merck Index: 12,08477 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...