Sodium-4-hydroxybenzoate 99,0

Supplier: TCI America

Synonyms: 4-Hydroxybenzoate sodium, Sodium 4-hydroxybenzoate, p-Hydroxybenzoic acid Monosodium Salt, 4-Hydroxybenzoic Acid Sodium Salt

H0221-25G H0221-500G
TCH0221-025GEA 88.48 CAD
TCH0221-025G TCH0221-500G
Sodium-4-hydroxybenzoate 99,0
Sodium-4-hydroxybenzoate

CAS Number: 114-63-6
MDL Number: MFCD00016530
Molecular Formula: C7H6O3
Molecular Weight: 160.10
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 300


Other Product Names / Synonyms: 4-Hydroxybenzoic Acid Sodium Salt

Formula: C₇H₆O₃
MW: 160.10 g/mol
Melting Pt: 300 °C
MDL Number: MFCD00016530
CAS Number: 114-63-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bb652fee52eebb31d9b810160411d58a74699ef1&vid=bb652fee52eebb31d9b810160411d58a74699ef1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR