Nonyl-4-hydroxybenzoate ≥98.0%

Supplier: TCI America

Synonyms: 4-Hydroxybenzoic Acid Nonyl Ester, Nonylparaben

H0217-500G H0217-25G H0217-100G
TCH0217-500GEA 1135.55 CAD
TCH0217-500G TCH0217-025G TCH0217-100G
Nonyl-4-hydroxybenzoate ≥98.0%
Nonyl-4-hydroxybenzoate

CAS Number: 38713-56-3
MDL Number: MFCD00016483
Molecular Formula: C16H24O3
Molecular Weight: 264.37
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 44


Other Product Names / Synonyms: 4-Hydroxybenzoic Acid Nonyl Ester, Nonylparaben

Formula: C₁₆H₂₄O₃
Melting Pt: 44 °C
MDL Number: MFCD00016483
CAS Number: 38713-56-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9dd2dc1886f506ab3033abdcbe609c1e17f832bd&vid=9dd2dc1886f506ab3033abdcbe609c1e17f832bd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR