Heptylnonyl adipate (so called)

Supplier: TCI America

Synonyms: Adipic Acid Heptylnonyl Ester

A1388-25ML A1388-500ML
TCA1388-025MLEA 36.34 CAD
TCA1388-025ML TCA1388-500ML
Heptylnonyl adipate (so called)
Heptylnonyl adipate

[Plasticizer]
MDL Number: MFCD00236062
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 0.92


Other Product Names / Synonyms: Adipic Acid Heptylnonyl Ester

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=512b852664a38e4a4a39136ae8145c0a5b9bdd5e&vid=dc4454559d5359f49933baf2d7d6888d5b6dffcf&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR