Acetaldehyde semicarbazone ≥98.0% (by titrimetric analysis)

Supplier: TCI America

A0193-5G A0193-25G
TCA0193-5GEA 75.84 CAD
TCA0193-5G TCA0193-025G
Acetaldehyde semicarbazone ≥98.0% (by titrimetric analysis)
Acetaldehyde semicarbazone

CAS Number: 591-86-6
MDL Number: MFCD00059191
Molecular Formula: C3H7N3O
Molecular Weight: 101.11
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 164

Formula: C₃H₇N₃O
Melting Pt: 164 °C
MDL Number: MFCD00059191
CAS Number: 591-86-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dc114b635eed5d28f6155c36825575422df67c19&vid=dc114b635eed5d28f6155c36825575422df67c19&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR