Diisononyl adipate (so called)
Supplier: TCI America
A1387-25ML
A1387-500ML
TCA1387-025MLEA
53.72
CAD
TCA1387-025ML
TCA1387-500ML
Diisononyl adipate (so called)
Diisononyl adipate
[Plasticizer]
MDL Number: MFCD00054165
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 232
Specific Gravity (20/20): 0.92
Other Product Names / Synonyms: Adipic Acid Diisononyl Ester
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...