DL-Histidine 98,0 for synthesis
Supplier: TCI America
Synonyms:
2-Amino-3-(1H-imidazol-4-yl)propanoic acid, H-DL-His-OH
H0148-25G
H0148-1G
TCH0148-025GEA
390.26
CAD
TCH0148-025G
TCH0148-001G
DL-Histidine 98,0 for synthesis
DL-Histidine
CAS Number: 4998-57-6
MDL Number: MFCD00005208
Molecular Formula: C6H9N3O2
Molecular Weight: 155.16
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 286
Product Categories: Bioscience, Chemical Mediators, Proteinogenic Amino Acids
Other Product Names / Synonyms: H-DL-His-OH
Formula:
C₆H₉N₃O₂ MW: 155.16 g/mol Melting Pt: 273 |
MDL Number:
MFCD00005208 CAS Number: 4998-57-6 EINECS: 206-513-8 Merck Index: 12,04758 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...