Hinokitiol ≥99.0%

Supplier: TCI America

Synonyms: beta-Thujaplicine, 2-Hydroxy-4-isopropylcyclohepta-2,4,6-trien-1-one

H0142-1G H0142-5G
TCH0142-001GEA 66.36 CAD
TCH0142-001G TCH0142-005G
Hinokitiol ≥99.0%
Hinokitiol

CAS Number: 499-44-5
MDL Number: MFCD00059582
Molecular Formula: C10H12O2
Molecular Weight: 164.20
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 138
Melting point (°C): 53
Flash Point (°C): 137


Product Categories: Bioscience, Monocyclic Monoterpenes
Other Product Names / Synonyms: 2-Hydroxy-4-isopropylcyclohepta-2,4,6-trien-1-one, beta-Thujaplicine

Formula: C₁₀H₁₂O₂
MW: 164.20 g/mol
Boiling Pt: 138 °C
Melting Pt: 53 °C
Flash Pt: 137 °C
MDL Number: MFCD00059582
CAS Number: 499-44-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b31e0a43151b07e89974597040455514b510a9a&vid=7b31e0a43151b07e89974597040455514b510a9a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR